missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Polyester Sign: CORROSIVE MATERIALS. WEAR REQUIRED PROTECTION.
Improve workplace safety with proper signage and markings. This Brady™ polyester sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$13.41 - $34.50
Specifications
| Material | Polyester |
|---|---|
| Shape | Rectangular |
| Displayed Text | CORROSIVE MATERIALS. WEAR REQUIRED PROTECTION. |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
|
19-106-450
|
Brady™
144950 |
14 x 10 in. |
Each for $34.50
|
|
|||||
|
19-106-452
|
Brady™
144952 |
10 x 7 in. |
Each for $13.41
|
|
|||||
Description
- Polyester material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
| Polyester | |
| Rectangular | |
| CORROSIVE MATERIALS. WEAR REQUIRED PROTECTION. |