missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Polyester Overlaminate for Labels
Description
- Clear overlaminate material available in three standard sizes to fit majority of existing Right-To-Know labels
- Standard package: 50 labels
Specifications
Specifications
| Product Type | Label Overlaminate |
| Color | Clear |
| For Use With (Equipment) | Brady Label Printers |
| Material | Polyester |
| Dimensions (L x W) | 10.5 x 7.5 in. (W x H) |
| Quantity | 50/Pk. |