Learn More
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 770
CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol
Supplier: Thermo Scientific Chemicals 042497AK
Description
Polydimethylsiloxane, trimethylsiloxy is used in coating Auxiliary Agents. It is also used as Pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Colorless | |
| M.W. 770 | |
| 9016-00-6 | |
| -40°C | |
| MFCD00084411 | |
| Soluble in aromatic hydrocarbon,partially soluble in aliphatic and insoluble in water. | |
| SEUDSDUUJXTXSV-UHFFFAOYSA-N | |
| dimethylsilanone | |
| 135°C (275°F) | |
| 1.397 |
| Polydimethylsiloxane, trimethylsiloxy terminated | |
| Liquid | |
| >205°C | |
| (C2H6OSi)n | |
| 0.918 g/mL | |
| 250 mL | |
| C[Si](C)(-*)O-* | |
| 74.15 | |
| Odorless |
Chemical Identifiers
| 9016-00-6 | |
| 74.15 | |
| SEUDSDUUJXTXSV-UHFFFAOYSA-N | |
| C[Si](C)(-*)O-* |
| (C2H6OSi)n | |
| MFCD00084411 | |
| dimethylsilanone |
Safety and Handling
RTECSNumber : TQ2690000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only