missing translation for 'onlineSavingsMsg'
Learn More

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 410

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol

Supplier:  Thermo Scientific Chemicals 043669AK

 View more versions of this product

Catalog No. AA43669AK

Only null left
Explore available promotions

Description

Description

Polydimethylsiloxane trimethylsiloxy terminated, is widely used as a surfactant, as a component of defoamers which are used to suppress the formation of foams. It is also used as a critical ingredient in water-repelling coatings. It is used as an active silicone fluid, as a stamp resin in the procedure of soft lithography, in optic telecommunications, in biomedical research, as elasticity of polymer networks, as adhesive materials and in skin-moisturizing lotions. It plays an important role in filler fluid in breast implants and as a sorbent for the analysis of headspace of food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Colorless
M.W. 410
9016-00-6
-40°C
MFCD00084411
Slightly miscible with water. Immiscible with methanol and ethylene glycol.
SEUDSDUUJXTXSV-UHFFFAOYSA-N
dimethylsilanone
70°C (158°F)
1.39
Polydimethylsiloxane, trimethylsiloxy terminated
Liquid
85°C to 105°C (2 mmHg to 4 mmHg)
(C2H6OSi)n
0.88 g/mL
250 mL
C[Si](C)(-*)O-*
74.15
Odorless

Chemical Identifiers

9016-00-6
74.15
SEUDSDUUJXTXSV-UHFFFAOYSA-N
C[Si](C)(-*)O-*
(C2H6OSi)n
MFCD00084411
dimethylsilanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TQ2690000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only