missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 117,000
CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol
Supplier: Thermo Scientific Chemicals 042504AK
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Colorless | |
| M.W. 117,000 | |
| 9016-00-6 | |
| <-40°C | |
| MFCD00084411 | |
| 250 mL | |
| C[Si](C)(-*)O-* | |
| 74.15 | |
| Odorless |
| Polydimethylsiloxane, trimethylsiloxy terminated | |
| Viscous Liquid | |
| >205°C | |
| (C2H6OSi)n | |
| 0.976 g/mL | |
| SEUDSDUUJXTXSV-UHFFFAOYSA-N | |
| dimethylsilanone | |
| 315°C (599°F) | |
| 1.4035 |
Chemical Identifiers
| 9016-00-6 | |
| 74.15 | |
| SEUDSDUUJXTXSV-UHFFFAOYSA-N | |
| C[Si](C)(-*)O-* |
| (C2H6OSi)n | |
| MFCD00084411 | |
| dimethylsilanone |
Safety and Handling
RTECSNumber : TQ2690000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only