missing translation for 'onlineSavingsMsg'
Learn More

Polydimethylsiloxane, Silanol Terminated, Spectrum™ Chemical

Catalog No. 18605294
Change view
Click to view available options
Quantity:
100 g
1 kg
3 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605294 100 g Amber Glass Bottle
18605295 1 kg Amber Glass Bottle
18605296 3 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18605294 Supplier Spectrum Chemical Mfg Cor Supplier No. P2952100GM
Get Up to $500 in Chemicals with Purchase
View all promotions

P2952, 70131-67-8

Spectrum™ Chemical Polydimethylsiloxane, Silanol Terminated is a silicon-based organic polymer with an average molecular weight of 70,000 - 80,000. It is used in cosmetic and personal care products as a surfactant and conditioning agent. It appears as a clear, viscous liquid and is insoluble in water.

Chemical Identifiers

CAS 70131-67-8
Molecular Formula HOSi
Molecular Weight (g/mol) 45.09
MDL Number MFCD01325012
InChI Key XQSFXFQDJCDXDT-UHFFFAOYSA-N
IUPAC Name hydroxy-λ¹-silanylidyne
SMILES O[Si]

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 100 g
Viscosity 15000-20000 cSt
Grade Reagent