missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PolyCo™ VR™ Protective Boot Covers
Description
- Knee-high cover with elastic top
- Two supple weights provide flexible, lightweight coverage
- Available in different colors
- Standard or cleanroom packaged (for Class 100 controlled environments)
Specifications
Specifications
| Size | Regular |
| Material | Polyolefin |
| Color | Blue |
| Closure Type | Elastic Top |
| Seam Type | Heat sealed |
| Thickness | 4 mil |
| For Use With (Application) | Ideal for use where biological and chemical liquids are present |