Learn More
PolyCo™ VR™ Protective Ankle-High Shoe Covers
Designed for comfort, flexibility and protection
Supplier: PolyCo™ 49515
Description
- Choice of 4mil or 6mil thickness
- Water fluid resistance
- Chemical resistance
- Elastic top for added splash protection
- Gritted material surface for slip resistance
- Made in USA
Specifications
| Large | |
| Yellow | |
| Heat sealed | |
| Ideal for use where biological and chemical liquids are present |
| Polyolefin | |
| Elastic Top | |
| 6 mil |