missing translation for 'onlineSavingsMsg'
Learn More

PolyCo™ PolyWear™ Smooth Gowns, Individually Bagged Bulk Packaged

Catalog No. 19125611
Encompass
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19125611 X-Large
19125610 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19125611 Supplier PolyCo™ Supplier No. 10610

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Smooth finish gowns provide lightweight impervious protection against biohazards.

  • In compliance with FDA requirements and tested in accordance with ASTM standards including Breaking Load, Tensile Strength and Elongation
  • Successfully tested for liquid and aerosol blood resistance under ASTM F 1670 Synthetic Blood Penetration Resistance and ASTM F 1671 Bacteriophage Penetration Resistance
  • In compliance with OSHA Bloodborne Pathogen Standard, 29, CFR Part 1910.1030
  • Meets AAMI Levels 1-4
  • Superior physical properties over imports
  • Easy, tear-away feature for quick gown removal and safe disposal
  • Contoured thumb loops secure gown sleeve for added protection
  • Redirects stress to the film material not the thermal seals to minimize breakage potential
  • Full-length open back affords better comfort and shoulder coverage
  • Provides liquid and aerosol blood resistance
  • Lightweight, non-linting construction

Specifications

For Use With (Application) Hospitals, Health Clinics, Laboratories
Gender Unisex
Product Type General Purpose Gown
No. of Pockets None
Packaging Type Bulk, Individually Bagged
Size X-Large
Material Polyethylene Film
Color Blue
Closure Type Over-the-Head
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Finish Smooth Matte
Latex Free Yes
Model 10610
Sleeve Style Full Sleeve with Thumb Loop
Certifications/Compliance ASTM F1670, ASTM F1671, AAMI Level 1 and 2, OSHA 29, CFR Part 1910.1030
Autoclavable Not Autoclavable
Show More Show Less