Learn More
Corning™ Polycarbonate Erlenmeyer Flask with Dip Tube, Male Luer Lock, Sterile
Provide optimum visibility and durability
$1216.25 - $2171.41
Specifications
Closure Color Orange
Closure Material Polypropylene
Material Polycarbonate
Disposable Single-use
Mouth Style Threaded
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Closure Includes Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Closure Includes Price Quantity  
15-100-150
View Documents
Corning™
C11405
125 mL 33 mm Vent, 1/8 in. dip tube, graduations
Case of 6 Each for $2,171.41
Only null left
15-100-151
View Documents
Corning™
11415
250 mL 38 mm 0.2μm Vent, 1/8 in. dip tube, male luer lock
Case of 4 Each for $1,469.12
Only null left
15-100-153
View Documents
Corning™
11465
2 L 48 mm 0.2μm Vent, 1/4 in. dip tube, male luer lock disconnect
Case of 2 Each for $1,216.25
Only null left
15-100-154
View Documents
Corning™
11495
3 L 70 mm 1/4 in. dip tube, male luer lock disconnect
Case of 2 Each for $1,318.98
Only null left
Description
Description
Corning™ closed system Erlenmeyer flasks will enhance the productivity of your shaker culture operations when adding or removing medium and cells. Flask is made from optically clear polycarbonate and is preassembled with an aseptic transfer cap. Unit is sterilized by gamma radiation and is nonpyrogenic and RNase-/DNase-free.
Flask:
  • Polycarbonate construction: USP Class VI material
  • Molded-in graduations for accuracy (+/-5%)
Aseptic Transfer Cap:
  • Chemically resistant, heat-sealable, heat-sealable, flexible tubing, 1/8” ID, 1/4” OD
  • Male Luer Lok Disconnect
Specifications
Specifications
Orange
Polycarbonate
Threaded
Polypropylene
Single-use
Aseptic Transfer Cap
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.