missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Polycarbonate Dome for FastPrep-24™
Polycarbonate Dome is a protective cover for FastPrep-24™ Classic bead beater.
Supplier: MP Biomedicals™ 116002506
Description
- Applications:
- Dome for FastPreps-24
- Polycarbonate protective cover
Specifications
| Polycarbonate | |
| 1 Ea. |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.