missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Polyacrylonitrile
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
$488.38 - $488.38
Specifications
| CAS | 25014-41-9 |
|---|---|
| Density | 1.184 g/mL (at 25°C) |
| Linear Formula | (C3H3N)n |
| Refractive Index | n20/D 1.514 |
| MDL Number | MFCD00084395 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
181315100G
|
Sigma Aldrich
181315100G |
100 g |
Each for $488.38
|
|
|||||
Description
PAN molecule has strong polar nitrile groups.It is relatively insoluble in nature. PAN based carbon fibers possess very high strength compared to other polymeric precursors. When subjected to heat treatment, it can produce high carbon yield giving rise to thermally stable, highly oriented, graphite like molecular structure. Generating carbon-fiber from PAN based fiber is a combination of three processes-- namely stabilization, carbonization and graphitization.Polyacrylonitrile (PAN) is used as polymeric carbon precursor to form carbon fibers, electrospun activated carbon materials having meso-macro pores, carbon black additives.These are consecutively used in hydrogen storage, EMI shielding, electrochemistry, separation processes. It may find applications in PAN based single walled carbon nanotube composites.Specifications
| 25014-41-9 | |
| (C3H3N)n | |
| MFCD00084395 | |
| 150000 |
| 1.184 g/mL (at 25°C) | |
| n20/D 1.514 | |
| Vinyl cyanide |