missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Polyacrylonitrile

Catalog No. 181315100G
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
181315100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 181315100G Supplier Sigma Aldrich Supplier No. 181315100G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

PAN molecule has strong polar nitrile groups.It is relatively insoluble in nature. PAN based carbon fibers possess very high strength compared to other polymeric precursors. When subjected to heat treatment, it can produce high carbon yield giving rise to thermally stable, highly oriented, graphite like molecular structure. Generating carbon-fiber from PAN based fiber is a combination of three processes-- namely stabilization, carbonization and graphitization.Polyacrylonitrile (PAN) is used as polymeric carbon precursor to form carbon fibers, electrospun activated carbon materials having meso-macro pores, carbon black additives.These are consecutively used in hydrogen storage, EMI shielding, electrochemistry, separation processes. It may find applications in PAN based single walled carbon nanotube composites.

Specifications

CAS 25014-41-9
Density 1.184 g/mL (at 25°C)
Refractive Index n20/D 1.514
Linear Formula (C3H3N)n
Quantity 100 g
MDL Number MFCD00084395
Synonym Vinyl cyanide
Molecular Weight (g/mol) 150000