missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Poly(vinylpolypyrrolidone), ∼110 μm

Catalog No. 111015780
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015780 Supplier MilliporeSigma™ Supelco™ Supplier No. 77627100G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Poly(vinylpolypyrrolidone) is a crosslinked homopolymer form of purified vinylpyrrolidone.

Poly(vinylpolypyrrolidone) is hygroscopic in nature. It has been used widely in plant hormone purification. It binds with phenolic compounds by hydrogen bonding.

Specifications

Particle Size 110 μm
Quantity 100 g
MDL Number MFCD00149016
CAS 9003-39-8