missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Poly(vinylpolypyrrolidone), ∼110 μm

Catalog No. 111015778
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015778 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015778 Supplier MilliporeSigma™ Supelco™ Supplier No. 77627500G

Poly(vinylpolypyrrolidone) is a crosslinked homopolymer form of purified vinylpyrrolidone.

Poly(vinylpolypyrrolidone) is hygroscopic in nature. It has been used widely in plant hormone purification. It binds with phenolic compounds by hydrogen bonding.

Specifications

Particle Size 110 μm
Quantity 500 g
MDL Number MFCD00149016
CAS 9003-39-8