missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl chloride), carboxylated, aver. M.W.220000, aver. carboxyl content 1.8 wt%

CAS: 25702-80-1 | (C2H3R)n | 134.56 g/mol

$249.09 - $249.09

Chemical Identifiers

CAS 25702-80-1
Molecular Formula (C2H3R)n
Molecular Weight (g/mol) 134.56
MDL Number MFCD00084460
InChI Key SQNNHEYXAJPPKH-UHFFFAOYSA-N
Synonym 2-propenoic acid, polymer with chloroethene, polyvinyl chloride-acrylic copolymer, acrylic acid; chloroethylene, chloroethene; prop-2-enoic acid, acrylic acid; vinyl chloride
PubChem CID 3080734
IUPAC Name chloroethene; prop-2-enoic acid
SMILES *-CC(-*)[*]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC183311000
SDS View Documents
Thermo Scientific Chemicals
183311000
100 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25702-80-1
134.56
SQNNHEYXAJPPKH-UHFFFAOYSA-N
3080734
*-CC(-*)[*]
(C2H3R)n
MFCD00084460
2-propenoic acid, polymer with chloroethene, polyvinyl chloride-acrylic copolymer, acrylic acid; chloroethylene, chloroethene; prop-2-enoic acid, acrylic acid; vinyl chloride
chloroethene; prop-2-enoic acid

Specifications

White
25702-80-1
MFCD00084460
2-propenoic acid, polymer with chloroethene, polyvinyl chloride-acrylic copolymer, acrylic acid; chloroethylene, chloroethene; prop-2-enoic acid, acrylic acid; vinyl chloride
SQNNHEYXAJPPKH-UHFFFAOYSA-N
chloroethene; prop-2-enoic acid
3080734
Glass bottle
Poly(vinyl chloride), carboxylated
Amorphous Powder
(C2H3R)n
1.3900g/mL
100 g
*-CC(-*)[*]
134.56
Authentic
1.39
Videos
SDS
Documents

Documents

RUO – Research Use Only