missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl acetate), approx. M.W. 100,000

Catalog No. AC183265000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC183265000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC183265000 Supplier Thermo Scientific Chemicals Supplier No. 183265000
Only null left

CAS: 9003-20-7 | (C4H6O2)n | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 9003-20-7
Molecular Formula (C4H6O2)n
Molecular Weight (g/mol) 86.09
MDL Number MFCD00084457
InChI Key XTXRWKRVRITETP-UHFFFAOYSA-N
Synonym vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
PubChem CID 7904
ChEBI CHEBI:46916
IUPAC Name ethenyl acetate
SMILES CC(=O)OC(-*)C-*

Specifications

Physical Form Beads
Linear Formula [-CH2CH(O2CCH3)-]n
Density 1.1800g/mL
Quantity 500 g
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Glass bottle
Chemical Name or Material Poly(vinyl acetate), Average M.W. 101600

RUO – Research Use Only