missing translation for 'onlineSavingsMsg'
Learn More

Pyramex Safety Products GL402C5 Series Polyurethane Palm-Coated Gloves

Catalog No. p-7182072
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Glove Cuff Color:
Black
Blue
Brown
Red
Yellow
Catalog No. Size Glove Cuff Color
19555054 Small Red
19555053 Medium Yellow
19555052 Large Brown
19555056 X-Large Black
19555055 2X-Large Blue
5 options

Catalog No. 19555054

Supplier: Pyramex Safety Products GL402C5S

Only null left

Polyurethane palm-coated gloves are tear and abrasion resistant, while offering tactile sensitivity and dexterity.

  • 13-gauge cut-resistant HPPE liner
  • Low particulate shed
  • Grips well without being sticky
  • Offers tactile sensitivity and dexterity
  • ANSI Cut Level: 4
  • Abrasion Rating: 4
  • Circular Blade Cut Rating: 5
  • Tear Rating: 4
  • Puncture Rating: 3

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material Polyurethane
Color Gray, Green
Cut Resistance Rating ANSI A4
Material Polyurethane
Product Type Cut-resistant Gloves
Size Small
EN388 Standard EN 388:4X43D
For Use With (Application) Electronics, general duty, parts assembly, sheet metal, handling, glass cutting
Glove Cuff Color Red
Glove Cuff Style Elasticated
Liner Material HPPE
Model GL402C5S
Puncture Rating ASTM F2878 LEVEL 3
Series GL402C5 Series Polyurethane Dipped
Show More Show Less