missing translation for 'onlineSavingsMsg'
Learn More

Pyramex Safety Products GL401 Polyurethane Coated Nylon Gloves

Catalog No. 19555049
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19555049 Small
19555048 Medium
19555047 Large
19555051 X-Large
19555050 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19555049 Supplier Pyramex Safety Products Supplier No. GL401S
Only null left

Gloves are designed to enhance grip capabilities without being sticky, in addition to providing a high degree of dexterity and tactile sensitivity.

  • 13 gauge nylon gloves with polyurethane coating
  • Provides dexterity and tactile sensitivity
  • Enhanced grip capabilities
  • Low particulate shed

Specifications

ANSI Abrasion Rating ANSI 1
Cuff Material Latex Elastic
Coating Material Polyurethane
Color Gray
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
EN388 Standard 4131X
For Use With (Application) Construction, Electronics, Inspection, and Quality Control
Glove Cuff Style Knit
Puncture Rating ANSI/ISEA 105-16 Level 2
Series GL401
Yarn Gauge 13
Show More Show Less