missing translation for 'onlineSavingsMsg'
Learn More

Poly(tetrafluoroethylene)

Catalog No. p-186522
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC178930050 5 g Glass Bottle
AC178931000 100 g Plastic bottle
AC178932500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC178930050 Supplier Thermo Scientific Chemicals Supplier No. 178930050
Only null left

CAS: 9002-84-0 | (C2F4)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 9002-84-0
Molecular Formula (C2F4)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084455
Synonym PTFE
SMILES FC(F)(-*)C(F)(F)-*

Specifications

Color White
Physical Form Powder
Linear Formula (-CF2CF2-)n
Density 2.0000g/mL
Quantity 5 g
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Glass Bottle
Chemical Name or Material Poly(tetrafluoroethylene)
RTECSNumber KX4025000
TSCA TSCA

RUO – Research Use Only