missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Poly(tetrafluoroethylene)
Description
Poly(tetrafluoroethylene) is a fluoropolymer that is commercially known as PTFE. Its properties include high thermal stability, excellent chemical resistance, low dielectric constant and low surface energy. It is a hydrophobic polymer that is majorly used as a protective coating on the metal surface.Inert lubricant.
Specifications
Specifications
| CAS | 9002-84-0 |
| Density | 2.15 g/mL (at 25°C) |
| Quantity | 100 g |
| Linear Formula | (CF2CF2)n |
| MDL Number | MFCD00084455 |
| Synonym | Fluorinated polymer; Fluoropolymer; PTFE |