missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Poly(tetrafluoroethylene)
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
$333.09 - $333.09
Specifications
| CAS | 9002-84-0 |
|---|---|
| Density | 2.15 g/mL (at 25°C) |
| Linear Formula | (CF2CF2)n |
| MDL Number | MFCD00084455 |
| Synonym | Fluorinated polymer; Fluoropolymer; PTFE |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
430935100G
|
Sigma Aldrich
430935100G |
100 g |
Each for $333.09
|
|
|||||
Description
Poly(tetrafluoroethylene) is a fluoropolymer that is commercially known as PTFE. Its properties include high thermal stability, excellent chemical resistance, low dielectric constant and low surface energy. It is a hydrophobic polymer that is majorly used as a protective coating on the metal surface.Inert lubricant.Specifications
| 9002-84-0 | |
| (CF2CF2)n | |
| Fluorinated polymer; Fluoropolymer; PTFE |
| 2.15 g/mL (at 25°C) | |
| MFCD00084455 |