missing translation for 'onlineSavingsMsg'
Learn More

Poly(styrene sulfonic acid) sodium salt, M.W. 70,000

Catalog No. AA4168830
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4168830 250 g
AA4168818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4168830 Supplier Thermo Scientific Chemicals Supplier No. 04168830

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25704-18-1 | (C8H7NaO3S)n | 184.21 g/mol

Poly(styrene sulfonic acid) sodium salt is used as Polyelectrolyte. It is also used as electroconductive and antistatic resin for electrographic and electrophotographic substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25704-18-1
Molecular Formula (C8H7NaO3S)n
Molecular Weight (g/mol) 184.21
MDL Number MFCD00084449
InChI Key MAGFQRLKWCCTQJ-UHFFFAOYSA-N
Synonym 4-styrenesulfonic acid sodium salt
PubChem CID 131675404
IUPAC Name 4-ethenylbenzenesulfonic acid;molecular hydrogen;sodium
SMILES [Na]OS(=O)(=O)C1=CC=C(C=C1)C(-*)C-*

Specifications

Quantity 250 g
Solubility Information Soluble in water,lower alcohols,glycols.
Physical Form Powder
Chemical Name or Material Poly(styrene sulfonic acid) sodium salt, M.W. 70,000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only