missing translation for 'onlineSavingsMsg'
Learn More

Poly(styrene-divinylbenzene), 1% cross-linked, 200-400 mesh

Catalog No. AA4180006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4180006 5 g
AA4180014 25 g
AA4180022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4180006 Supplier Thermo Scientific Chemicals Supplier No. 4180006

For the preparation of functionalized beads used in solid-phase synthesis | CAS: 9003-70-7 | C24H27NO5 | 409.48 g/mol

Poly(styrene-divinylbenzene) is widely used as column packing material in liquid chromatography for separation of both organic and inorganic ions. is used in inorganic trace analysis for solid phase extraction preconcentration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9003-70-7
Molecular Formula C24H27NO5
Molecular Weight (g/mol) 409.48
MDL Number 9003-70-7
InChI Key BCIPGSZQUDLGSY-UHFFFAOYNA-N
Synonym styrene/divinylbenzen, divinylbenzene-styrene, styrene divinylbenzene, styrene-divinylbenzene, styrene/divinylbenzene, styrene-divinyl benzene, benzene, diethenyl-, polymer with ethenylbenzene, brominated, st dvb, divinylbenzene; styrene, 1,2-diethenylbenzene; styrene
PubChem CID 174664
IUPAC Name 1,2-bis(ethenyl)benzene;styrene
SMILES CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C=O

Specifications

Physical Form Beads
Solubility Information Insoluble in water.
Quantity 5 g
Mesh Size 200 to 400 Mesh
Odor Odorless
Chemical Name or Material Poly(styrene-divinylbenzene), 1% cross-linked
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only