missing translation for 'onlineSavingsMsg'
Learn More

Poly(sodium-p-styrenesulfonate), average M.W. 70.000

Catalog No. AC222271000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC222271000 100 g Plastic bottle
AC222270050 5 g Glass Bottle
AC222275000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC222271000 Supplier Thermo Scientific Chemicals Supplier No. 222271000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25704-18-1 | (C8H7NaO3S)n | 184.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25704-18-1
Molecular Formula (C8H7NaO3S)n
Molecular Weight (g/mol) 184.21
MDL Number MFCD00084449
InChI Key MAGFQRLKWCCTQJ-UHFFFAOYSA-N
Synonym 4-styrenesulfonic acid sodium salt
PubChem CID 131675404
SMILES [Na]OS(=O)(=O)C1=CC=C(C=C1)C(-*)C-*

Specifications

Color Tan to White
Physical Form Powder
Quantity 100 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula [-CH2CH(C6H4SO3Na)-]n
Solubility Information Solubility in water: soluble. Other solubilities: soluble in lower glycerols
IUPAC Name 4-ethenylbenzenesulfonic acid;molecular hydrogen;sodium
Chemical Name or Material Poly(sodium-p-styrenesulfonate), Average M.W. 70.000
RTECSNumber DB8550000
TSCA TSCA

RUO – Research Use Only