missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Poly Sign: DANGER: H2S POIS. GAS MAY BE PRESENT. NO OPEN FLAMES. WEAR NECES. PROTECT. EQUIP AND PROCEED w/ CAUTION.

Catalog No. 19106538
Change view
Click to view available options
Dimensions (L x W):
10 x 7 in.
14 x 10 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19106538 14 x 10 in.
19106537 10 x 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19106538 Supplier Brady™ Supplier No. 145067

Improve workplace safety with proper signage and markings. This Brady™ polystyrene sign is a durable, clear and effective tool to raise awareness to workplace hazards.

  • Polystyrene material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Polystyrene signs are thin, pliable and durable
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use

Specifications

Material Polystyrene
Shape Rectangular
Dimensions (L x W) 14 x 10 in.
Displayed Text DANGER: H2S POISONOUS GAS MAY BE PRESENT. NO OPEN FLAMES. WEAR NECESSARY PROTECTIVE EQUIPMENT AND PROCEED WITH CAUTION.
Color Blue