missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StatLab Poly Scientific Sodium Chloride 0.85% Aqueous
Description
- Sodium Chloride 0.85% Aqueous solution formulated to maintain cell viability and structural integrity, making it suitable for use throughout histological and laboratory workflows.
- Commonly used as a carrier for bacterial suspensions, helping to preserve cell viability during processing and analysis.
- Also utilised in the preparation of buffer solutions to maintain appropriate osmotic pressure and pH for cell culture and a range of cell analysis applications.
- Supplied as a StatLab Poly Scientific Special Stain Kit component, ensuring consistent quality, compatibility, and reliable laboratory performance.
Specifications
Specifications
| Physical Form | Aqueous Solution |
| CAS | 67-56-1, 64-17-5, 67-63-0, 64-19-7, 50-00-0 |
| Color | Clear Colorless |
| Quantity | 2.5 gal. |
| Grade | Laboratory |