missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StatLab Poly Scientific Chromic Acid 10% Aqueous
Description
- Chromic Acid 10% Aqueous is primarily used as a fixative to preserve cell structures and tissue samples for microscopic examination
- The fixative function of chromic acid is based on its ability to cross-link proteins and other cellular components, resulting in the stabilization of cellular structures
- A StatLab - Poly Scientific Special Stain Kit Component
Spécifications
Spécifications
| Nom ou substance chimique | Chromic Acid |
| Concentration | 10% |
| Quantité | 16 oz. |