Learn More
StatLab Poly Scientific Ammonia Water
Description
- It is a strong alkaline solution that helps to blue sections adequately, making it easier to see the details of the tissue samples
- The pH of ammonia water is typically around 10, which is considered too high for some procedures, however, it is still used in some labs as a last resort to ensure sections do not wash off or wrinkle
- It is important to note that ammonia water can cause tissue to fall off slides and is a strong oxidizer and corrosive, therefore, it should be handled with care and under proper ventilation and safety measures
- A StatLab - Poly Scientific Special Stain Kit Component
- 4 gallons per case
Specifications
Specifications
| Type | Ammonia Water |
| Quantity | 4 x 1 gal. |
| Form | Liquid |