missing translation for 'onlineSavingsMsg'
Learn More

Poly(propylene glycol), average M.W. 4.000

Catalog No. p-186454
Change view
Click to view available options
Quantity:
250 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC192202500 250 g Glass bottle
AC192205000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC192202500 Supplier Thermo Scientific Chemicals Supplier No. 192202500
Only null left

CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25322-69-4
Molecular Formula (C3H6O)n
Molecular Weight (g/mol) 134.18
MDL Number MFCD00084446
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
PubChem CID 32881
IUPAC Name 2-(2-hydroxypropoxy)propan-1-ol
SMILES [H]OC(C)CO

Specifications

Color Green to Yellow
Physical Form Viscous Liquid
Linear Formula H[OCH(CH3)CH2]nOH
Density 1.0000g/mL
Quantity 250 g
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4500 to 1.4520
Specific Gravity 1
Viscosity 930 mm2/s (25°C)
Chemical Name or Material Poly(propylene glycol), Average M.W. 4.000
Show More Show Less
RTECSNumber OE5784360
TSCA TSCA

RUO – Research Use Only