missing translation for 'onlineSavingsMsg'
Learn More

Poly(propylene glycol), average M.W. 2.000

Catalog No. p-186429
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC192185000 500 g Glass bottle
AC192180025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC192185000 Supplier Thermo Scientific Chemicals Supplier No. 192185000

CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25322-69-4
Molecular Formula (C3H6O)n
Molecular Weight (g/mol) 134.18
MDL Number MFCD00084446
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
PubChem CID 32881
IUPAC Name 2-(2-hydroxypropoxy)propan-1-ol
SMILES [H]OC(C)CO

Specifications

Color White to Gray
Physical Form Crystals or Crystalline Powder
Acid Value 0.1 mg KOH/g max.
Boiling Point 200.0°C
Melting Point -31.0°C
Linear Formula H[OCH(CH3)CH2]nOH
Density 1.0000g/mL
Solubility Information Solubility in water: practically insoluble.
Quantity 500 g
Flash Point 230°C
Hydroxyl Value 53 to 58 mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4495 to 1.4515
Specific Gravity 1
Viscosity 300 mm2/s (25°C)
Water 0.15% Max.
Chemical Name or Material Poly(propylene glycol), Average M.W. 2.000
Show More Show Less
RTECSNumber OE5784360
TSCA TSCA

RUO – Research Use Only