missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly[[N-isopropylacrylamide-co-[N-[3-(dodecyldimethylammonio)propyl]acrylamide]]phosphotungstate], TCI America™
$288.94 - $956.25
Specifications
| Color | Yellow |
|---|---|
| Quantity | 200 mg |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Poly[[N-isopropylacrylamide-co-[N-[3-(dodecyldimethylammonio)propyl]acrylamide]]phosphotungstate] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P1697200MG
|
TCI America
P1697200MG |
200 mg |
Each for $288.94
|
|
|||||
|
P16971G
|
TCI America
P16971G |
1 g |
Each for $956.25
|
|
|||||
Specifications
| Yellow | |
| 200 mg | |
| Crystalline Powder | |
| Poly[[N-isopropylacrylamide-co-[N-[3-(dodecyldimethylammonio)propyl]acrylamide]]phosphotungstate] |
Safety and Handling
TSCA : No