missing translation for 'onlineSavingsMsg'
Learn More

Poly(methylhydrosiloxane)

Catalog No. AAL1456118
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1456118 50 g
AAL1456130 250 g
AAL145610B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1456118 Supplier Thermo Scientific Chemicals Supplier No. L1456118

CAS: 63148-57-2 | (CH4OSi)n

Poly(methylhydrosiloxane) is used in the reduction of esters to alcohols, catalyzed by a combination of titanocene dichloride and either n-BuLi or EtMgBr. It is a safer alternative to triethoxysilane, B22063, for reduction of phosphine oxides to phosphines, catalyzed by Ti(O-i-Pr)4. It is also used in greener amine synthesis by reductive amination as an alternative to borohydrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63148-57-2
Molecular Formula (CH4OSi)n
MDL Number MFCD00084478
InChI Key QNWOFLWXQGHSRH-UHFFFAOYSA-N
Synonym Methyl hydrogen polysiloxane; PMHS
SMILES C[SiH](O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Density 1.006
Solubility Information Not miscible or difficult to mix in water.
Quantity 50 g
Formula Weight ca 1900
Flash Point 204°C (399°F)
Refractive Index 1.398
Chemical Name or Material Poly(methylhydrosiloxane)
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only