missing translation for 'onlineSavingsMsg'
Learn More

Poly(methyl methacrylate), approx. M.W. 15000 (GPC)

Catalog No. p-186380
Change view
Click to view available options
Quantity:
50 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC190690500 50 g Plastic bottle
AC190691000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC190690500 Supplier Thermo Scientific Chemicals Supplier No. 190690500

CAS: 9011-14-7 | (C5H8O2)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9011-14-7
Molecular Formula (C5H8O2)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00134349
InChI Key PMAMJWJDBDSDHV-UHFFFAOYSA-N
Synonym Methyl methacrylate, polymerized, PMMA
IUPAC Name 1-methoxy-2-methyl-1-oxopropan-2-yl
SMILES COC(=O)C(C)(-*)C-*

Specifications

Color White
Physical Form Beads
Linear Formula [-CH2C(CH3)(CO2CH3)-]n
Density 1.18
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, chloroform, mek, thf
Quantity 50 g
Flash Point >250°C
Infrared Spectrum Authentic
Loss on Drying 3% max.
Packaging Plastic bottle
Particle Size Bead size (typically): 10 to 100 micron
Chemical Name or Material Poly(methyl methacrylate), Average M.W. 15000 (GPC)
Show More Show Less
RTECSNumber TR0400000
TSCA TSCA

RUO – Research Use Only