missing translation for 'onlineSavingsMsg'
Learn More

Poly(methyl methacrylate), For GPC, average Mw 97,000 (Typical), average Mn 46,000 (Typical), MilliporeSigma™ Supelco™

Catalog No. 111015833
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015833 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015833 Supplier Merck Emd Millipore Supplier No. 37003725G

Chemical Identifiers

MDL Number MFCD00134349
Synonym PMMA; Poly(methacrylic acid methyl ester)

Specifications

Physical Form Neat
CAS 9011-14-7
Linear Formula [CH2 C(CH3)(CO2 CH3)]n
Density 1.200 g/cm3
Solubility Information Water: insoluble
Quantity 25 g
Flash Point >250°C
Grade GPC
Packaging Poly Bottle
RTECSNumber TR0400000