Learn More
Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 2,400
Fluorosilicone lubricant | CAS: 63148-56-1 | C4H7F3OSi | 156.179 g/mol
$152.18 - $526.05
Chemical Identifiers
| CAS | 63148-56-1 |
|---|---|
| Molecular Formula | C4H7F3OSi |
| Molecular Weight (g/mol) | 156.179 |
| MDL Number | MFCD00241477 |
| InChI Key | CUMCLKHSZZGALD-UHFFFAOYSA-N |
| Synonym | methyl 3,3,3-trifluoropropyl silanone, poly methyl-3,3,3-trifluoropropylsiloxane, methyl-oxo-3,3,3-trifluoropropyl silane, 5,5,5-trifluoro-2-silapentane-2-one, poly 3,3,3-trifluoropropylmethylsiloxane, methyl 3,3,3-trifluoropropyl cyclopolysiloxanes, silane, methyloxo 3,3,3-trifluoropropyl, methyl 3,3,3-trifluoropropyl cyclopolysiloxanes, mixture of tri-and tetra |
| PubChem CID | 4285173 |
| IUPAC Name | methyl-oxo-(3,3,3-trifluoropropyl)silane |
| SMILES | C[Si](=O)CCC(F)(F)F |
Description
Fluorosilicone lubricantPoly(methyl-3,3,3-trifluoropropylsiloxane) is used in the preparation of siliconized- glass fiber sheets, which acts as a support for protein-chemical analysis of electroblotted proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 63148-56-1 | |
| 156.179 | |
| CUMCLKHSZZGALD-UHFFFAOYSA-N | |
| 4285173 | |
| C[Si](=O)CCC(F)(F)F |
| C4H7F3OSi | |
| MFCD00241477 | |
| methyl 3,3,3-trifluoropropyl silanone, poly methyl-3,3,3-trifluoropropylsiloxane, methyl-oxo-3,3,3-trifluoropropyl silane, 5,5,5-trifluoro-2-silapentane-2-one, poly 3,3,3-trifluoropropylmethylsiloxane, methyl 3,3,3-trifluoropropyl cyclopolysiloxanes, silane, methyloxo 3,3,3-trifluoropropyl, methyl 3,3,3-trifluoropropyl cyclopolysiloxanes, mixture of tri-and tetra | |
| methyl-oxo-(3,3,3-trifluoropropyl)silane |
Specifications
| Colorless | |
| 63148-56-1 | |
| MFCD00241477 | |
| methyl 3,3,3-trifluoropropyl silanone, poly methyl-3,3,3-trifluoropropylsiloxane, methyl-oxo-3,3,3-trifluoropropyl silane, 5,5,5-trifluoro-2-silapentane-2-one, poly 3,3,3-trifluoropropylmethylsiloxane, methyl 3,3,3-trifluoropropyl cyclopolysiloxanes, silane, methyloxo 3,3,3-trifluoropropyl, methyl 3,3,3-trifluoropropyl cyclopolysiloxanes, mixture of tri-and tetra | |
| 25 g | |
| C[Si](=O)CCC(F)(F)F | |
| 156.179 | |
| 260°C (500°F) | |
| Poly (methyl-3,3,3-trifluoropropylsiloxane), M.W. 2,400 |
| Liquid | |
| C4H7F3OSi | |
| 1.25 g/mL | |
| Immiscible with water. | |
| CUMCLKHSZZGALD-UHFFFAOYSA-N | |
| methyl-oxo-(3,3,3-trifluoropropyl)silane | |
| 4285173 | |
| 1.381 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only