missing translation for 'onlineSavingsMsg'
Learn More

Poly(methyl-3,3,3-trifluoropropylsiloxane), M.W. 2,400

Catalog No. AA4322814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4322814 25 g
AA4322822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4322814 Supplier Thermo Scientific Chemicals Supplier No. 04322814
Only null left

Fluorosilicone lubricant | CAS: 63148-56-1 | C4H7F3OSi | 156.179 g/mol

Fluorosilicone lubricantPoly(methyl-3,3,3-trifluoropropylsiloxane) is used in the preparation of siliconized- glass fiber sheets, which acts as a support for protein-chemical analysis of electroblotted proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 63148-56-1
Molecular Formula C4H7F3OSi
Molecular Weight (g/mol) 156.179
MDL Number MFCD00241477
InChI Key CUMCLKHSZZGALD-UHFFFAOYSA-N
Synonym methyl 3,3,3-trifluoropropyl silanone, poly methyl-3,3,3-trifluoropropylsiloxane, methyl-oxo-3,3,3-trifluoropropyl silane, 5,5,5-trifluoro-2-silapentane-2-one, poly 3,3,3-trifluoropropylmethylsiloxane, methyl 3,3,3-trifluoropropyl cyclopolysiloxanes, silane, methyloxo 3,3,3-trifluoropropyl, methyl 3,3,3-trifluoropropyl cyclopolysiloxanes, mixture of tri-and tetra
PubChem CID 4285173
IUPAC Name methyl-oxo-(3,3,3-trifluoropropyl)silane
SMILES C[Si](=O)CCC(F)(F)F

Specifications

Color Colorless
Physical Form Liquid
Density 1.25 g/mL
Solubility Information Immiscible with water.
Quantity 25 g
Flash Point 260°C (500°F)
Refractive Index 1.381
Chemical Name or Material Poly (methyl-3,3,3-trifluoropropylsiloxane), M.W. 2,400
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only