missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Poly(I)·Poly(C)
Description
- For stimulation of interferon production in vitro
- Supplied as a lyophilized powder
- Induces interferon in rabbit cells
- May be effective for inducing human interferon in vitro
Specifications
Specifications
| Quantity | 265 mg |
| Color | White |
| For Use With (Application) | Activation of Various Cell Types |
| Form | Powder |