missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Poly(ethyleneimine) solution

Catalog No. 181978250G
Encompass
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
181978250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 181978250G Supplier Sigma Aldrich Supplier No. 181978250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Protein precipitant.

Specifications

CAS 9002-98-6
Quantity 250 g
Synonym Ethyleneimine polymer solution; PEI
Molecular Weight (g/mol) 750000