missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(ethylene oxide), M.W. 100,000 to 200,000
CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
$477.17 - $477.17
Chemical Identifiers
| CAS | 25322-68-3 |
|---|---|
| Molecular Formula | (C2H4O)n |
| Molecular Weight (g/mol) | 62.07 |
| MDL Number | MFCD01779601 |
| InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
| Synonym | PEO |
| IUPAC Name | ethane-1,2-diol |
| SMILES | [H]OCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC178585000
|
Thermo Scientific Chemicals
178585000 |
500 g | Glass bottle |
Each for $477.17
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 25322-68-3 | |
| 62.07 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol |
| (C2H4O)n | |
| MFCD01779601 | |
| PEO | |
| [H]OCCO |
Specifications
| White to Yellow | |
| 25322-68-3 | |
| (C2H4O)n | |
| MFCD01779601 | |
| PEO | |
| 500 g | |
| [H]OCCO | |
| 62.07 | |
| 1% max. | |
| 1.4539 |
| Powder | |
| 65.0°C to 67.0°C | |
| (-CH2CH2O-)n | |
| 15, 7688 | |
| Solubility in water: soluble. Other solubilities: soluble in many organic solvents | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| Authentic | |
| Glass bottle | |
| Poly(ethylene oxide), Approximate M.W. 100.000 |
Safety and Handling
RTECSNumber : TQ4100000
TSCA : TSCA
RUO – Research Use Only