missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(ethylene glycol methyl ether), average M.W. 750
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| pH | 4 to 7 (5% aq. soln.) |
| Physical Form | Fused Mass |
| Avg. Mol. Wt. or Mol. Wt. Range | 715 to 785 |
| Linear Formula | CH3(OCH2CH)nOH |
| Density | 1.09g/mL |
| Quantity | 500 g |
| IUPAC Name | 2-methoxyethanol |
| Hydroxyl Value | 71.5 to 78.5mg KOH/g |
| Infrared Spectrum | Authentic |
| Show More |
Safety and Handling
| RTECSNumber | PE8263000 |
| TSCA | TSCA |
RUO – Research Use Only