missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Poly(ethylene glycol) diacrylate

Catalog No. 455008500ML
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
455008500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 455008500ML Supplier Sigma Aldrich Supplier No. 455008500ML
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(ethylene glycol) diacrylate

Specifications

CAS 26570-48-9
Melting Point 12°C to 17°C
Density 1.12 g/mL (at 25°C)
Refractive Index n20/D 1.47
MDL Number MFCD00081876
Quantity 500 mL
Synonym PEG diacrylate
Molecular Weight (g/mol) 700