missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol), average M.W. 4000

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

$293.93 - $293.93

Chemical Identifiers

Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEO
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC434630010
SDS View Documents
Thermo Scientific Chemicals
434630010
1 kg Plastic Bottle
Each for $293.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

(C2H4O)n
MFCD01779601
PEO
[H]OCCO
62.07
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol

Specifications

White
54.0°C to 58.0°C
H(OCH2CH2)nOH
15,7688
1 kg
[H]OCCO
62.07
Authentic
0.5% max.
Flakes
(C2H4O)n
MFCD01779601
PEO
LYCAIKOWRPUZTN-UHFFFAOYSA-N
ethane-1,2-diol
4000
Plastic Bottle
Poly(ethylene glycol)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 500-038-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only