missing translation for 'onlineSavingsMsg'
Learn More

Poly(ethylene glycol), average M.W. 300

Catalog No. p-3737757
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC192220010 1 L Glass bottle
AC192220025 2.5 L Glass bottle
AC192220100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC192220010 Supplier Thermo Scientific Chemicals Supplier No. 192220010
Only null left

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25322-68-3
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 62.07
MDL Number MFCD01779601
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym PEG
IUPAC Name ethane-1,2-diol
SMILES [H]OCCO

Specifications

Color Clear liquid
Melting Point -15.0°C to +10.0°C
Linear Formula H(OCH2CH2)nOH
Density 1.1200g/mL
Merck Index 15, 7688
Solubility Information Solubility in water: soluble. Other solubilities: soluble in many organic solvents
Quantity 1 L
Flash Point >180°C
Hydroxyl Value 356 to 394mg KOH/g
Packaging Glass bottle
Refractive Index 1.4620 to 1.4650
Specific Gravity 1.12
Viscosity 85-100 mPa.s (20°C)
Chemical Name or Material Poly(ethylene glycol)
Show More Show Less
EINECSNumber 500-038-2

RUO – Research Use Only