missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CritiCore Protective Wear Poly Cotton Lab Coat in White
Poly cotton lab coat in white designed with 3 pockets, knit cuff sleeves, and a snap front enclosure.
Supplier: CritiCore Protective Wear 12L2505382PC3P3XLWHT
Description
- 80/20 Poly Cotton
- Knit Cuff Sleeves
- Snap Front Enclosure
- 3 Exterior Pockets
- 1 Pencil Slot Lapel
- Collar Side Vents
- Primary Applications: Medical / Lab
Specifications
| 3X-Large | |
| Polycotton | |
| Snaps | |
| Reusable | |
| Particulates | |
| 3 |
| Unisex | |
| White | |
| Knit | |
| Non-sterile | |
| Regular |