missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(Bisphenol A carbonate), Molecular Weight Series, MilliporeSigma™ Supelco™
Description
Poly(Bisphenol A carbonate) (PC) is a thermoplastic material. It is injected in molding operations and is majorly studied to understand its thermal behaviour, especially thermal degradation property. It can be synthesized by inducing supercritical CO2 to crystallinity in low molecular weight polycarbonate beads using solid-state polymerization method.
Specifications
Specifications
| CAS | 25037-45-0 |
| Density | 1.2 g/mL (at 25°C) |
| Purity Grade Notes | A lot specific analysis is included with each unit |
| Physical Form | Neat |
| MDL Number | MFCD00084476 |
| Type | Analytical Standard |
| Quantity | 1 Ea. |
| Grade | Analytical Standard |