missing translation for 'onlineSavingsMsg'
Learn More

Poly(Allylamine Hydrochloride)

Catalog No. AA4309209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4309209 10 g
AA4309218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4309209 Supplier Thermo Scientific Chemicals Supplier No. 04309209

As a polymeric primary amine; Forms hydrogels | CAS: 71550-12-4 | C3H7N | 93.56 g/mol

Poly(allylamine hydrochloride) is used in film and polymer industries and cell encapsulation. Further, it is used to make redox hydrogel- modified electrodes for measuring enzyme responses. In addition, it is used in the preparation of an antireflective coating consists of alternate layers of poly(allyamine hydrochloride) and silica nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71550-12-4
Molecular Formula C3H7N
Molecular Weight (g/mol) 93.56
MDL Number MFCD00054329
InChI Key VVJKKWFAADXIJK-UHFFFAOYSA-N
Synonym allylamine hydrochloride, 3-aminopropene hydrochloride, poly allylamine hydrochloride, allylamine, hydrochloride, allylammonium chloride, 2-propen-1-amine, hydrochloride, unii-r2h25ilf98, 2-propen-1-amine hydrochloride, allylaminehydrochloride, prop-2-en-1-amine hydrochloride
PubChem CID 82291
IUPAC Name prop-2-en-1-amine;hydrochloride
SMILES NCC=C

Specifications

Color Yellow
Physical Form Powder
Solubility Information Soluble in water.
Quantity 10 g
Chemical Name or Material Poly (Allylamine Hydrochloride)
Hazard Category H302-H317
Hazard Statement GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
EINECSNumber 415-050-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only