Learn More
Enzo Life Sciences Poly(ADP-ribose) monoclonal antibody (10H) (100 l)
Supplier: Enzo ALX804220R100
Clone: 10H. Host: Mouse. Isotype: IgG3. Immunogen: Purified poly(ADP-ribose). Species Reactivity: Human, Mouse, Rat, Drosophila. Specificity: Recognizes poly(ADP-ribose) synthesized by a broad range of PARPs (poly(ADP-ribose) polymerases) like human, mouse, rat or Drosophila PARP enzyme. Applications: Flow Cytometry, ICC, IHC (PS), WB. Recommended dilutions: Immunocytochemistry (5-20g/ml). Immunohistochemistry (paraffin sections; dilution buffer: 5% milk (non fat dried milk) in PBS to a final. Concentration of 5-20g/ml). Western Blot (incubate 2.5g/ml in PBS, 0.05% Tween20, 5% milk (non fat dried milk)). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A-affinity purified from supernatant. Formulation: Liquid. In 50mM HEPES, pH 7.4,containing 100mM sodium chloride, 1% BSA and 0.02% sodium azide. Handling: Avoid freeze/thaw cycles.