missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly(acrylic acid), sec. stand., aver. MW 1,080,000, aver. MN 135,000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Linear Formula | [-CH2CH(CO2H)-]n |
| Solubility Information | Solubility in water: soluble. |
| Quantity | 1 g |
| IUPAC Name | prop-2-enoic acid |
| Formula Weight | 1.080.000 (Average ) |
| Infrared Spectrum | Authentic |
| Loss on Drying | 10% max. |
| Packaging | Glass bottle |
| Show More |
Safety and Handling
| RTECSNumber | AT4680000 |
| TSCA | TSCA |
RUO – Research Use Only