missing translation for 'onlineSavingsMsg'
Learn More

Merck Emd Millipore poly(A) | 26763-19-9 | MFCD00131937 | 1g

Supplier:  Merck Emd Millipore P94031G

Encompass

poly(A) | 26763-19-9 | MFCD00131937 | 1g

Catalog No. 501779522


Only null left